SY3H3D67723A Synthonix Corporation

6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one

  • MDL number MFCD08689590
  • Cas Number : 21594-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41837.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41837.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart