SY3H3D677254 Synthonix Corporation

1-Bromo-2-chloro-4-nitrobenzene

  • MDL number MFCD00051514
  • Cas Number : 29682-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Yellow - Black
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41909.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41909.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart