SY3H3D677279 Synthonix Corporation

5-Bromo-3-(trifluoromethyl)pyridin-2-ol

  • MDL number MFCD12546486
  • Cas Number : 76041-79-7
  • Molecular Weight : 242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42003.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42003.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart