SY3H3D67727F Synthonix Corporation

2-Bromo-4-chloro-6-fluorophenol

  • MDL number No MDL found
  • Cas Number : 886499-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42047.html

Safety Information

MSDS https://synthonix.com/42047.html
Availability details for
Quantity
  • Availability
  • Add to Cart