SY3H3D677280 Synthonix Corporation

2-Bromo-6-chloro-4-fluorophenol

  • MDL number MFCD04115954
  • Cas Number : 886499-83-8
  • Molecular Weight : 225.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42048.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42048.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart