SY3H3D67728D Synthonix Corporation

2-Bromo-4-chloro-6-nitrophenylamine

  • MDL number MFCD08275709
  • Cas Number : 827-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white to yellow to orange powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42090.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42090.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart