SY3H3D67728F Synthonix Corporation

3-Bromo-2-chloro-6-methyl-5-nitropyridine

  • MDL number No MDL found
  • Cas Number : 856834-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42092.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42092.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart