SY3H3D677290 Synthonix Corporation

3-Bromo-6-chloro-2-methyl-5-nitropyridine

  • MDL number MFCD09839274
  • Cas Number : 186413-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown to Brown Solid or Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42093.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42093.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart