SY3H3D6772B6 Synthonix Corporation

2-Bromo-3,4-difluoroaniline

  • MDL number MFCD11035924
  • Cas Number : 1092349-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Off-white to gray to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42209.html

Safety Information

signalword Warning
pcodes P261, P280, P405
MSDS https://synthonix.com/42209.html
pictograms GHS07
hstatements H302, H312, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart