SY3H3D6772D4 Synthonix Corporation

3-Bromo-4-chlorophenol

  • MDL number MFCD00070740
  • Cas Number : 13659-24-0
  • Molecular Weight : 207.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to Yellow to Brown or Pale-red Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42340.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42340.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart