SY3H3D6772DA Synthonix Corporation

4-Bromo-3-fluorophenol

  • MDL number MFCD00051907
  • Cas Number : 121219-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Off white to gray to yellow or pale-red powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42346.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42346.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart