SY3H3D6772E2 Synthonix Corporation

2-Bromo-4-nitroaniline

  • MDL number MFCD00025152
  • Cas Number : 13296-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Very pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42381.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42381.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart