SY3H3D6772E4 Synthonix Corporation

2-Bromo-6-nitroaniline

  • MDL number MFCD09263476
  • Cas Number : 59255-95-7
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown to Red Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42383.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42383.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart