SY3H3D6772E6 Synthonix Corporation

3-Bromo-4-nitroaniline

  • MDL number MFCD07783047
  • Cas Number : 40787-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Yellow to dark-brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42385.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42385.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart