SY3H3D6772EF Synthonix Corporation

2-((5-Bromopyrimidin-2-yl)thio)acetic acid

  • MDL number No MDL found
  • Cas Number : 52767-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42397.html

Safety Information

MSDS https://synthonix.com/42397.html
Availability details for
Quantity
  • Availability
  • Add to Cart