SY3H3D677306 Synthonix Corporation

3-Bromo-5-chlorophenylamine

  • MDL number MFCD06797804
  • Cas Number : 96558-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42527.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42527.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart