SY3H3D677314 Synthonix Corporation

2-Bromo-3-fluoroaniline

  • MDL number MFCD07369915
  • Cas Number : 111721-75-6
  • Molecular Weight : 190.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to brown to black or light-green to green to brown solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42541.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42541.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart