SY3H3D677316 Synthonix Corporation

3-Bromo-5-fluoroaniline

  • MDL number MFCD07779529
  • Cas Number : 134168-97-1
  • Molecular Weight : 190.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow to Purple to Brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42543.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42543.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart