SY3H3D677318 Synthonix Corporation

2-Bromo-3-fluoro-4-picoline

  • MDL number MFCD04112499
  • Cas Number : 884494-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to Yellow Liquid or Semi-Solid or solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42545.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42545.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart