SY3H3D67731B Synthonix Corporation

2-Bromo-5-fluoro-4-methylpyridine

  • MDL number MFCD08277310
  • Cas Number : 885168-20-7
  • Molecular Weight : 190.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/42548.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42548.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart