SY3H3D677337 Synthonix Corporation

5-Bromo-3-fluoro-1-methylpyridin-2(1H)-one

  • MDL number MFCD23135714
  • Cas Number : 1352152-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42656.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42656.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart