SY3H3D67733D Synthonix Corporation

(3-Bromo-2-fluorophenyl)boronic acid

  • MDL number MFCD05664228
  • Cas Number : 352535-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42687.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42687.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart