SY3H3D67735E Synthonix Corporation

2-Bromo-4-(methylsulfonyl)pyridine

  • MDL number MFCD28402467
  • Cas Number : 1193244-93-7
  • Molecular Weight : 236.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/42858.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42858.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart