SY3H3D67736B Synthonix Corporation

6-Bromo-1-methylpyridin-2(1H)-one

  • MDL number MFCD00234174
  • Cas Number : 873383-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42892.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42892.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart