SY3H3D67738C Synthonix Corporation

2-Bromo-4-methoxy-6-methylpyrimidine

  • MDL number MFCD28741601
  • Cas Number : 56545-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43058.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43058.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart