SY3H3D677393 Synthonix Corporation

5-Bromo-N,N-dimethylpyrazin-2-amine

  • MDL number MFCD09879029
  • Cas Number : 446286-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43265.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43265.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart