SY3H3D67739D Synthonix Corporation

4-Bromo-1-isopropylpyrazole

  • MDL number MFCD09801021
  • Cas Number : 313735-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43323.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43323.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart