SY3H3D6773A1 Synthonix Corporation

2-(tert-Butylamino)acetic acid hydrochloride

  • MDL number MFCD03788441
  • Cas Number : 6939-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43496.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43496.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart