SY3H3D6773AD Synthonix Corporation

(S)-4-Isobutyloxazolidine-2,5-dione

  • MDL number MFCD03411292
  • Cas Number : 3190-70-3
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43777.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43777.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart