SY3H3D6773B8 Synthonix Corporation

2-(2-Bromoethoxy)tetrahydro-2H-pyran

  • MDL number MFCD01321310
  • Cas Number : 17739-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Clear liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43864.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43864.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart