SY3H3D6773E6 Synthonix Corporation

1-Bromo-4-iodo-2-(trifluoromethyl)benzene

  • MDL number MFCD11855999
  • Cas Number : 364-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44025.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44025.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart