SY3H3D6773E8 Synthonix Corporation

4-Bromo-2-iodo-1-(trifluoromethyl)benzene

  • MDL number MFCD21603817
  • Cas Number : 1256945-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Colorless to Light-red to Orange Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44027.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44027.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart