SY3H3D6773EA Synthonix Corporation

2-Bromo-6-fluorobenzotrifluoride

  • MDL number MFCD01631569
  • Cas Number : 261951-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44029.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44029.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart