SY3H3D6773EE Synthonix Corporation

2-Bromo-1-fluoro-3-(trifluoromethyl)benzene

  • MDL number MFCD00061243
  • Cas Number : 104540-42-3
  • Molecular Weight : 243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow - Purple
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/44033.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44033.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart