SY3H3D6773F7 Synthonix Corporation

3-Bromo-2-(trifluoromethyl)imidazo[1,2-b]pyridazine

  • MDL number MFCD13193556
  • Cas Number : 1260817-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44083.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44083.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart