SY3H3D6773FB Synthonix Corporation

3-Bromo-4-chlorobenzonitrile

  • MDL number MFCD09025661
  • Cas Number : 948549-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white to pale-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44093.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44093.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart