SY3H3D6773FF Synthonix Corporation

5-Bromo-2-chlorobenzonitrile

  • MDL number MFCD00672948
  • Cas Number : 57381-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44097.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44097.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart