SY3H3D67741B Synthonix Corporation

4-Bromo-5-fluoro-2-nitrobenzoic acid

  • MDL number MFCD08702773
  • Cas Number : 1020717-99-0
  • Molecular Weight : 264.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Off white to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44169.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44169.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart