SY3H3D67741E Synthonix Corporation

1-(6-Bromopyridin-3-yl)-2,2,2-trifluoroethanone hydrate

  • MDL number MFCD28016685
  • Cas Number : 1956366-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44179.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44179.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart