SY3H3D67742A Synthonix Corporation

4-Bromo-2,6-difluorobenzoic acid

  • MDL number MFCD03094085
  • Cas Number : 183065-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44202.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44202.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart