SY3H3D677444 Synthonix Corporation

4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene

  • MDL number MFCD01861132
  • Cas Number : 105529-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44255.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44255.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart