SY3H3D677451 Synthonix Corporation

6-Bromo-8-chloroimidazo[1,2-a]pyridine

  • MDL number MFCD11518983
  • Cas Number : 474708-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Pale yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44301.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44301.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart