SY3H3D67747F Synthonix Corporation

5-Bromo-2-nitrobenzaldehyde

  • MDL number MFCD00456508
  • Cas Number : 20357-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44464.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44464.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart