SY3H3D6774B3 Synthonix Corporation

5-Bromo-2-chlorobenzaldehyde

  • MDL number MFCD08445659
  • Cas Number : 189628-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44642.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44642.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart