SY3H3D6774B7 Synthonix Corporation

2-Bromo-4-(trifluoromethyl)phenol

  • MDL number MFCD06657954
  • Cas Number : 81107-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44646.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44646.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart