SY3H3D6774D1 Synthonix Corporation

1-(Bromomethyl)-2,3-dichlorobenzene

  • MDL number MFCD03701317
  • Cas Number : 57915-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Lump - liquid Colour: White - Slightly pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44706.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44706.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart