SY3H3D6774D8 Synthonix Corporation

1-(Bromomethyl)-2-chloro-3-fluorobenzene

  • MDL number MFCD09038462
  • Cas Number : 874285-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Colorless to White to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44713.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44713.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart