SY3H3D6774D9 Synthonix Corporation

1-(Bromomethyl)-2-chloro-4-fluorobenzene

  • MDL number MFCD00236025
  • Cas Number : 45767-66-6
  • Molecular Weight : 223.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Very pale yellow - Pale reddish yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/44714.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44714.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart