SY3H3D6774E8 Synthonix Corporation

2-Bromo-5-cyano-4-picoline

  • MDL number MFCD09839265
  • Cas Number : 1003711-35-0
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to Yellow or Light-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44736.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44736.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart