SY3H3D677512 Synthonix Corporation

2-bromo-3-(trifluoromethyl)aniline

  • MDL number MFCD01656033
  • Cas Number : 58458-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to yellow to red solid or semi-solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44899.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44899.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart